Merck
  • Characterisation of testicular function and spermatogenesis in transgender women.
Characterisation of testicular function and spermatogenesis in transgender women.
Human reproduction (Oxford, England) (2020-12-02)
Gertjan Vereecke, Justine Defreyne, Dorien Van Saen, Sarah Collet, Jo Van Dorpe, Guy T'Sjoen, Ellen Goossens
ABSTRACT
Does gender-affirming treatment prevent full spermatogenesis in transgender women (TW)? Adequate hormonal therapy (HT) leads to complete suppression of spermatogenesis in most TW, if serum testosterone levels within female reference ranges are obtained. Gender-affirming treatment in transgender individuals may involve gender-affirming HT. The effects on spermatogenesis in TW remain unclear. In order to add information from a referral centre for transgender care, we wish to compare results of earlier studies with our population of TW who received a standard hormone treatment. This was a prospective cohort study part of the European Network for the Investigation of Gender Incongruence (ENIGI), conducted between 15 February 2010 and 30 September 2015. There were 162 TW were included in the ENIGI study at the Ghent University Hospital in Belgium. Participants are included in ENIGI when they first start HT, and follow-up visits occur over the next 3 years. The study included 97 TW who initiated HT with cyproterone acetate (CPA) plus oestrogens and proceeded with gonadectomy at the Ghent University Hospital. Testicular tissue retrieved during gonadectomy was processed and stained for four different germ cell markers by the Biology of the Testis lab at the Vrije Universiteit Brussel. Subsequent immunohistochemical staining was performed for melanoma-associated antigen A4 (MAGE-A4, marker for spermatogonia and early spermatocytes), boule homologue, RNA-binding protein (BOLL, marker for secondary spermatocytes and round spermatids), cAMP-responsive element modulator (CREM, marker for round spermatids) and acrosin (marker for acrosome visualization). Serum levels of sex steroids were measured prior to surgery. Suppressed testosterone levels (<50 ng/dl) were found in 92% of the participants prior to surgery. The mean time between initiation of HT and surgery was 685 days. In 88% (85/97) of the sections, MAGE-A4 staining was positive. Further staining could not reveal complete spermatogenesis in any participant. Testicular function of the participants prior to initiation of HT was not assessed, although all participants presented with cisgender male serum testosterone values before initiation of HT. The current study only reports on people using CPA at a fixed dose and may therefore not be applicable to all TW. HT leads to complete suppression of spermatogenesis in most TW, if serum testosterone levels within female reference ranges are obtained. Serum testosterone levels are associated with the sperm maturation rate. It is important to discuss sperm preservation before the start of hormone therapy. If serum testosterone levels remain higher, spermatogenesis may still occur. D.V.S. is a post-doctoral fellow of the Fonds Wetenschappelijk Onderzoek (FWO; 12M2819N). Processing of the testis specimens was funded by the Biology of The Testes (BITE) research group (Department of Reproduction, Genetics and Regenerative medicine at Vrije Universiteit Brussel (VUB)). There are no competing interests. N/A.
Sigma-Aldrich
Anti-CREM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.