Merck
  • Distinguishing the cyanobacterial neurotoxin β-N-methylamino-L-alanine (BMAA) from other diamino acids.
Distinguishing the cyanobacterial neurotoxin β-N-methylamino-L-alanine (BMAA) from other diamino acids.
Toxicon : official journal of the International Society on Toxinology (2011-02-19)
S A Banack, J S Metcalf, Z Spáčil, T G Downing, S Downing, A Long, P B Nunn, P A Cox
ABSTRACT
β-N-methylamino-L-alanine (BMAA) is produced by diverse taxa of cyanobacteria, and has been detected by many investigators who have searched for it in cyanobacterial blooms, cultures and collections. Although BMAA is distinguishable from proteinogenic amino acids and its isomer 2,4-DAB using standard chromatographic and mass spectroscopy techniques routinely used for the analysis of amino acids, we studied whether BMAA could be reliably distinguished from other diamino acids, particularly 2,6-diaminopimelic acid which has been isolated from the cell walls of many bacterial species. We used HPLC-FD, UHPLC-UV, UHPLC-MS, and triple quadrupole tandem mass spectrometry (UHPLC-MS/MS) to differentiate BMAA from the diamino acids 2,6-diaminopimelic acid, N-2(amino)ethylglycine, lysine, ornithine, 2,4-diaminosuccinic acid, homocystine, cystine, tryptophan, as well as other amino acids including asparagine, glutamine, and methionine methylsulfonium.
Sigma-Aldrich
L-2,4-Diaminobutyric acid dihydrochloride, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
2,6-Diaminopimelic acid, ≥98% (TLC)
Pricing and availability is not currently available.