Merck
  • Energetics and structural elucidation of mechanisms for gas phase H/D exchange of protonated peptides.
Energetics and structural elucidation of mechanisms for gas phase H/D exchange of protonated peptides.
The journal of physical chemistry. A (2010-10-26)
Blake E Ziegler, Terry B McMahon
ABSTRACT
Hydrogen/deuterium exchange reactions involving protonated triglycine and deuterated ammonia (ND(3)) have been examined in the gas phase using a Fourier transform ion cyclotron resonance (FT-ICR) mass spectrometer. Ab initio and density functional theory (DFT) calculations have been carried out to model the exchanges and to obtain energetics and vibrational frequencies for molecules involved in the proposed exchange mechanisms. Structural optimization and frequency calculations have been performed at the B3LYP level of theory with the 6-311+G(d,p) basis set. Transition states have been calculated at the same level of theory and basis set as above using the QST2 and QST3 methods. Single-point energy calculations have been performed at the MP2/6-311+G(d,p) level. Six labile sites of protonated triglycine were found to undergo H/D exchange. Of these six labile hydrogens, two are amide, three are ammonium, and one is carboxyl. Detailed mechanisms for each of these transfers are proposed. Qualitative onium ion and tautomer mechanisms for the exchanges of ammonium and amide hydrogens, respectively, using semiempirical calculations were suggested in previous studies by Beauchamp et al. As shown by the current ab initio and DFT calculations completed during this study, the mechanisms proposed in that study are notionally correct; however, the tautomer mechanisms are shown here to be the result of the fact that a second stable isomer of protonated triglycine exists in which the amide1 carbonyl oxygen is protonated. The exchange of the carboxyl hydrogen is found to proceed via a transition state resembling an ammonium ion interacting with a carboxylate moiety via two hydrogen bonds. The current work thus provides significant mechanistic and structural detail for a considerably more in-depth understanding of the processes involved in gas phase H/D exchange of peptides.
Sigma-Aldrich
Gly-Gly-Gly
Pricing and availability is not currently available.