Merck
Smurf2 exerts neuroprotective effects on cerebral ischemic injury.
The Journal of biological chemistry (2021-03-17)
Haibin Liu, Shengtao Sun, Bing Liu
ABSTRACT
The present study aimed to explore specific mechanisms involved in mediating the neuroprotective effects of Smad ubiquitination regulatory factor 2 (Smurf2) in cerebral ischemic injury. A middle cerebral artery occlusion (MCAO) mouse model and an oxygen-glucose deprivation (OGD)-treated neuron model were developed. The expression of Smurf2, Yin Yang 1 (YY1), hypoxia-inducible factor-1 alpha (HIF1α), and DNA damage-inducible transcript 4 gene (DDIT4) was analyzed. Thereafter, the expression of Smurf2, YY1, HIF1α, and DDIT4 was altered in the MCAO mice and OGD-treated neurons. Apoptosis in tissues and cerebral infarction were assessed. In neurons, the expression of apoptosis-related proteins, viability, and apoptosis were assessed, followed by evaluation of lactate dehydrogenase leakage rate. The interaction between Smurf2 and YY1 was analyzed by coimmunoprecipitation assay and that between YY1 ubiquitination by in vivo ubiquitination experiment. The results showed downregulation of Smurf2 and upregulation of YY1, HIF1α, and DDIT4 in both MCAO mice and OGD-treated neurons. Smurf2 elevated YY1 ubiquitination and degradation, and YY1 increased HIF1α expression to promote DDIT4 in neurons. Overexpressed Smurf2 or downregulated YY1, HIF1α, or DDIT4 reduced the volume of cerebral infarction and apoptosis in MCAO mice, while enhancing cell viability and reducing apoptosis and lactate dehydrogenase leakage in OGD-treated neurons. In summary, our findings elucidated a neuroprotective role of Smurf2 in cerebral ischemic injury via inactivation of the YY1/HIF1α/DDIT4 axis.
Product Description
Roche
Cytotoxicity Detection KitPLUS (LDH), sufficient for 400 multiwell tests (04744926001), sufficient for 2,000 multiwell tests (04744934001), kit of 1 (4 components), suitable for protein quantification, suitable for cell analysis, detection
Pricing and availability is not currently available.