Merck
  • Synthesis of aspartame precursor with an immobilized thermolysin in mixed organic solvents.
Synthesis of aspartame precursor with an immobilized thermolysin in mixed organic solvents.
Biotechnology and bioengineering (1995-06-20)
M Miyanaga, T Tanaka, T Sakiyama, K Nakanishi
ABSTRACT
N-(benzyloxycarbonyl)-L-aspartyl-L-phenylalanine methyl ester, a precursor of the synthetic sweetener, aspartame, was synthesized from N-(benzyloxycarbonyl)-L-aspartic acid and L-phenylalanine methyl ester with an immobilized thermolysin (EC 3.4.24.4) in the mixed organic solvent system of tert-amyl alcohol and ethyl acetate. A mixed solvent consisting of tert-amyl alcohol and ethyl acetate at a ratio of 33:67 (v/v) was found to be the most suitable with respect to synthetic rate and stability of the immobilized enzyme. The reaction continued to proceed quite successfully in a column reactor at 40 degrees C and at a space velocity of 3.6 h(-1) with a yield of 99%, using 40 mM Z-Asp and 200 mM PheOMe dissolved in the mixed solvent as the substrate. (c) 1995 John Wiley & Sons, Inc.
Sigma-Aldrich
Thermolysin from Geobacillus stearothermophilus, Type X, lyophilized powder, 30-350 units/mg protein (E1%/280)
Pricing and availability is not currently available.