Merck
  • Analysis of Static Molecular Gradients in a High-Throughput Drug Screening Microfluidic Assay.
Analysis of Static Molecular Gradients in a High-Throughput Drug Screening Microfluidic Assay.
Molecules (Basel, Switzerland) (2021-11-14)
Roman G Szafran, Benita Wiatrak
ABSTRACT
In this study, we thoroughly analyzed molecular gradient generation, its stability over time, and linearity in our high-throughput drug screening microfluidic assay (HTS). These parameters greatly affect the precision and accuracy of the device's analytical protocol. As part of the research, we developed a mathematical model of dependence of the concentration profile on the initial concentrations of active substances in reservoirs and the number of tilts, as well as the dependence of the active substance concentration profiles in the culture chambers on the concentration profile of the reference dye in the indicator chamber. The mean concentration prediction error of the proposed equations ranged from 1.4% to 2.4% for the optimized parameters of the procedure and did not increase with the incubation time. The concentration profile linearity index, Pearson's correlation coefficient reached -0.997 for 25 device tilts. The observed time stability of the profiles was very good. The mean difference between the concentration profile after 5 days of incubation and the baseline profile was only 7.0%. The newly created mathematical relationships became part of the new HTS biochip operating protocols, which are detailed in the article.
Product Description
Roche
Collagen, from rat tail tendon
Pricing and availability is not currently available.