Merck
  • Occurrence and fate of pharmaceutical and personal care products in a sewage treatment works.
Occurrence and fate of pharmaceutical and personal care products in a sewage treatment works.
Journal of environmental monitoring : JEM (2010-11-09)
Rubén Reif, Ana Santos, Simon J Judd, Juan M Lema, Francisco Omil
ABSTRACT
The occurrence and fate of eight pharmaceutical and personal care products (PPCPs) during sewage treatment has been studied in a pilot-scale treatment plant, comprising a primary settler (2.85 m(3)), an aeration tank (1.845 m(3)) and a secondary clarifier (0.5 m(3)), placed on site at a wastewater treatment works in the north west of the UK. It was fed both with raw sewage and the return liquor produced after sludge centrifugation, thus representing the most common configuration for a municipal sewage treatment plant based on the activated sludge process. Samples were taken at six different locations, including the return liquor stream, and analysed for musk fragrances and pharmaceutically active compounds belonging to various therapeutic groups such as anti-inflammatory drugs, tranquillisers and antiepileptics. Mass balances were conducted for those PPCPs that were quantifiable. The fate of the PPCPs was found to differ according to their physical-chemical characteristics. Anti-inflammatories underwent a degradation process and were almost completely removed from sewage during the biological treatment step. Musk fragrances were only partially removed, through adsorption onto the primary suspended solids and the biomass in the aerobic process, due to their strong lipophilic characteristics. The results of this study provide increasing evidence that the partial removal of these substances through the sewage treatment process contribute to the environmental occurrence of PPCPs. Consequently, existing STPs should be upgraded in order to attenuate the release of these substances into the aquatic environment.
Product Description
Millipore
Glass Fiber Membrane Filter, 1.2 μm Pore Size, Millipore, filter diam. 47 mm, hydrophilic
Pricing and availability is not currently available.