Merck
  • Transgenic potato expressing Abeta reduce Abeta burden in Alzheimer's disease mouse model.
Transgenic potato expressing Abeta reduce Abeta burden in Alzheimer's disease mouse model.
FEBS letters (2005-11-29)
Jung Won Youm, Hee Kim, Jee Hye Lo Han, Chang Hwan Jang, Hee Jin Ha, Inhee Mook-Jung, Jae Heung Jeon, Cheol Yong Choi, Young Ho Kim, Hyun Soon Kim, Hyouk Joung
ABSTRACT
Beta amyloid (Abeta) is believed one of the major pathogens of Alzheimer's disease (AD), and the reduction of Abeta is considered a primary therapeutic target. Immunization with Abeta can reduce Abeta burden and pathological features in transgenic AD model mice. Transgenic potato plants were made using genes encoding 5 tandem repeats of Abeta1-42 peptides with an ER retention signal. Amyloid precursor protein transgenic mice (Tg2576) fed with transgenic potato tubers with adjuvant showed a primary immune response and a partial reduction of Abeta burden in the brain. Thus, Abeta tandem repeats can be expressed in transgenic potato plants to form immunologically functional Abeta, and these potatoes has a potential to be used for the prevention and treatment of AD.
Sigma-Aldrich
Cholera Toxin B subunit, ≥95% (SDS-PAGE), lyophilized powder
Pricing and availability is not currently available.