Merck
  • Effects of 17β-Estradiol on growth-related genes expression in female and male spotted scat (Scatophagus argus).
Effects of 17β-Estradiol on growth-related genes expression in female and male spotted scat (Scatophagus argus).
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2020-09-06)
Xiao-Ying Ru, Hong-Juan Shi, Tuo Wang, Qian-Qing Liu, Dong-Neng Jiang, Yu-Hao Peng, Yang Huang, Chun-Hua Zhu, Shui-Sheng Li, Ran-Ran Dong, Guang-Li Li
ABSTRACT
Growth hormone (GH) is the most important endocrine factor to regulate somatic growth. Spotted scat (Scatophagus argus) is a famous marine aquaculture species in China with a typical sexual growth dimorphism in which females grow faster and larger than males. In this study, gh messenger RNA (gh mRNA) and GH protein expression were examined in the pituitary glands of female and male spotted scat. Based on qPCR analysis, gh mRNA was mainly expressed in the pituitary gland, and weakly in the gonads and hypothalamus. Furthermore, gh mRNA expression in the pituitary gland was significantly higher in females at stages II-IV than in males at stages III-V. In addition, gh mRNA was highly expressed in the ovary and testis during mature development stages. In this study, spotted scat GH polyclonal antibody was produced. Western blot analysis showed that the molecular weight of spotted scat GH was about 21 KDa. Immunohistochemistry (IHC) in pituitary glands showed that GH was mainly expressed in the proximal pars distal (PPD) and a few cells were distributed in the rostral pairs distal (RPD). After injecting 17β-Estradiol (E2) in vivo, gh mRNA expression was significantly up-regulated in the pituitary gland, whereas igf1 and ghr1 mRNA levels were down-regulated in the liver, which might regulate gh mRNA expression in the pituitary gland. These results provide valuable insight into the molecular mechanisms of E2 regulating gh expression in spotted scat.
Sigma-Aldrich
17β-Estradiol-16,16,17-d3, 98 atom % D, 99% (CP)
Pricing and availability is not currently available.