Merck
  • An Organic Borate Salt with Superior p-Doping Capability for Organic Semiconductors.
An Organic Borate Salt with Superior p-Doping Capability for Organic Semiconductors.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2020-10-01)
Berthold Wegner, Dominique Lungwitz, Ahmed E Mansour, Claudia E Tait, Naoki Tanaka, Tianshu Zhai, Steffen Duhm, Michael Forster, Jan Behrends, Yoshiaki Shoji, Andreas Opitz, Ullrich Scherf, Emil J W List-Kratochvil, Takanori Fukushima, Norbert Koch
ABSTRACT
Molecular doping allows enhancement and precise control of electrical properties of organic semiconductors, and is thus of central technological relevance for organic (opto-) electronics. Beyond single-component molecular electron acceptors and donors, organic salts have recently emerged as a promising class of dopants. However, the pertinent fundamental understanding of doping mechanisms and doping capabilities is limited. Here, the unique capabilities of the salt consisting of a borinium cation (Mes2B+; Mes: mesitylene) and the tetrakis(penta-fluorophenyl)borate anion [B(C6F5)4]- is demonstrated as p-type dopant for polymer semiconductors. With a range of experimental methods, the doping mechanism is identified to comprise electron transfer from the polymer to Mes2B+, and the positive charge on the polymer is stabilized by [B(C6F5)4]-. Notably, the former salt cation leaves during processing and is not present in films. The anion [B(C6F5)4]- even enables the stabilization of polarons and bipolarons in poly(3-hexylthiophene), not yet achieved with other molecular dopants. From doping studies with high ionization energy polymer semiconductors, the effective electron affinity of Mes2B+[B(C6F5)4]- is estimated to be an impressive 5.9 eV. This significantly extends the parameter space for doping of polymer semiconductors.
Sigma-Aldrich
Tris(pentafluorophenyl)borane, 95%
Pricing and availability is not currently available.