Merck
  • Metformin Mitigates DPP-4 Inhibitor-Induced Breast Cancer Metastasis via Suppression of mTOR Signaling.
Metformin Mitigates DPP-4 Inhibitor-Induced Breast Cancer Metastasis via Suppression of mTOR Signaling.
Molecular cancer research : MCR (2020-10-01)
Emi Kawakita, Fan Yang, Asako Kumagai, Yuta Takagaki, Munehiro Kitada, Yasuo Yoshitomi, Takayuki Ikeda, Yuka Nakamura, Yasuhito Ishigaki, Keizo Kanasaki, Daisuke Koya
ABSTRACT
The biological influence of antidiabetic drugs on cancer cells and diabetic cancer patients has not yet been completely elucidated. We reported that a dipeptidyl peptidase (DPP)-4 inhibitor accelerates mammary cancer metastasis by inducing epithelial-mesenchymal transition (EMT) through the CXCL12/CXCR4/mTOR axis. Metformin has been shown to inhibit the mTOR signaling pathway. In this study, we investigated whether metformin mitigates breast cancer metastasis induced by a DPP-4 inhibitor via suppression of mTOR signaling. In cultured mouse mammary and human breast cancer cells, metformin suppressed DPP-4 inhibitor KR62436 (KR)-induced EMT and cell migration via suppression of the mTOR pathway associated with AMPK activation. For the in vivo study, metformin intervention was performed in an allograft 4T1 breast cancer model mouse with or without KR. We also analyzed mice transplanted with shRNA-mediated DPP-4 knockdown 4T1 cells. Treatment with metformin inhibited the lung metastasis of DPP-4-deficient 4T1 mammary tumor cells generated by either KR administration or DPP-4 knockdown. Immunostaining of primary tumors indicated that DPP-4 suppression promoted the expression of EMT-inducing transcription factor Snail through activation of the CXCR4-mediated mTOR/p70S6K pathway in an allograft breast cancer model; metformin abolished this alteration. Metformin treatment did not alter DPP-4-deficiency-induced expression of CXCL12 in either plasma or primary tumors. Our findings suggest that metformin may serve as an antimetastatic agent by mitigating the undesirable effects of DPP-4 inhibitors in patients with certain cancers. IMPLICATIONS: Metformin could combat the detrimental effects of DPP-4 inhibitor on breast cancer metastasis via mTOR suppression, suggesting the potential clinical relevance. VISUAL OVERVIEW: http://mcr.aacrjournals.org/content/molcanres/19/1/61/F1.large.jpg.
Sigma-Aldrich
AMD3100 octahydrochloride hydrate, ≥97% (NMR), solid
Pricing and availability is not currently available.
Metformin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.