Merck
  • Injectable oligomer-cross-linked gelatine hydrogels via anhydride-amine-conjugation.
Injectable oligomer-cross-linked gelatine hydrogels via anhydride-amine-conjugation.
Journal of materials chemistry. B (2021-02-23)
Hafiz Awais Nawaz, Kathleen Schröck, Maximilian Schmid, Jan Krieghoff, Iram Maqsood, Christian Kascholke, Caroline Kohn-Polster, Michaela Schulz-Siegmund, Michael C Hacker
ABSTRACT
Injectable gelatine-based hydrogels are valuable tools for drug and cell delivery due to their extracellular matrix-like properties that can be adjusted by the degree of cross-linking. We have established anhydride-containing oligomers for the cross-linking of gelatine via anhydride-amine-conjugation. So far, this conversion required conditions not compatible with cell encapsulation or in vivo injection. In order to overcome this limitation, we developed an array of quarter-oligomers varying in comonomer composition and contents of reactive anhydride units reactive towards amine groups under physiological conditions. The oligomers were of low molecular weight (Mn < 5 kDa) with a high degree of chemically intact anhydrides. Chemical comonomer composition was determined by 1H-NMR. Dissolutions experiments confirmed improved hydrophilicity of the synthesized oligomers over our established compositions. Injectable formulations are described utilizing cytocompatible concentrations of constituent materials and proton-scavenging base. Degree of cross-linking and stiffness of injectable hydrogels were controlled by composition. The gels hold promise as injectable drug or cell carrier and as bioink.
Sigma-Aldrich
4-Acryloylmorpholine, 97%, contains 1,000 ppm monomethyl ether hydroquinone as inhibitor
Pricing and availability is not currently available.
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Pricing and availability is not currently available.