Merck
  • A potent HDAC inhibitor, 1-alaninechlamydocin, from a Tolypocladium sp. induces G2/M cell cycle arrest and apoptosis in MIA PaCa-2 cells.
A potent HDAC inhibitor, 1-alaninechlamydocin, from a Tolypocladium sp. induces G2/M cell cycle arrest and apoptosis in MIA PaCa-2 cells.
Journal of natural products (2014-07-08)
Lin Du, April L Risinger, Jarrod B King, Douglas R Powell, Robert H Cichewicz
ABSTRACT
The cyclic tetrapeptide 1-alaninechlamydocin was purified from a Great Lakes-derived fungal isolate identified as a Tolypocladium sp. Although the planar structure was previously described, a detailed analysis of its spectroscopic data and biological activity are reported here for the first time. Its absolute configuration was determined using a combination of spectroscopic ((1)H-(1)H ROESY, ECD, and X-ray diffraction) and chemical (Marfey's analysis) methods. 1-Alaninechlamydocin showed potent antiproliferative/cytotoxic activities in a human pancreatic cancer cell line (MIA PaCa-2) at low-nanomolar concentrations (GI50 5.3 nM, TGI 8.8 nM, LC50 22 nM). Further analysis revealed that 1-alaninechlamydocin induced G2/M cell cycle arrest and apoptosis. Similar to other cyclic epoxytetrapeptides, the inhibitory effects of 1-alaninechlamydocin are proposed to be produced primarily via inhibition of histone deacetylase (HDAC) activity.
Sigma-Aldrich
Histone Deacetylase Assay Kit, Fluorometric, sufficient for 100 assays (96 well plates)
Pricing and availability is not currently available.