Merck
  • Ephrin-A5/EphA4 signalling controls specific afferent targeting to cochlear hair cells.
Ephrin-A5/EphA4 signalling controls specific afferent targeting to cochlear hair cells.
Nature communications (2013-02-07)
Jean Defourny, Anne-Lise Poirrier, François Lallemend, Susana Mateo Sánchez, Jakob Neef, Pierre Vanderhaeghen, Eduardo Soriano, Christiane Peuckert, Klas Kullander, Bernd Fritzsch, Laurent Nguyen, Gustave Moonen, Tobias Moser, Brigitte Malgrange
ABSTRACT
Hearing requires an optimal afferent innervation of sensory hair cells by spiral ganglion neurons in the cochlea. Here we report that complementary expression of ephrin-A5 in hair cells and EphA4 receptor among spiral ganglion neuron populations controls the targeting of type I and type II afferent fibres to inner and outer hair cells, respectively. In the absence of ephrin-A5 or EphA4 forward signalling, a subset of type I projections aberrantly overshoot the inner hair cell layer and invade the outer hair cell area. Lack of type I afferent synapses impairs neurotransmission from inner hair cells to the auditory nerve. By contrast, radial shift of type I projections coincides with a gain of presynaptic ribbons that could enhance the afferent signalling from outer hair cells. Ephexin-1, cofilin and myosin light chain kinase act downstream of EphA4 to induce type I spiral ganglion neuron growth cone collapse. Our findings constitute the first identification of an Eph/ephrin-mediated mutual repulsion mechanism responsible for specific sorting of auditory projections in the cochlea.
Sigma-Aldrich
Anti-Neurofilament NF-H Antibody, clone TA51, culture supernatant, clone TA51, Chemicon®
Pricing and availability is not currently available.