Merck
  • Comparison of the binding affinity of CGP-12177A at recombinant rat alpha(1D)-adrenoceptors expressed in BHK-21 cell membranes and alpha(1)-adrenoceptors present in rat cerebral cortex membranes.
Comparison of the binding affinity of CGP-12177A at recombinant rat alpha(1D)-adrenoceptors expressed in BHK-21 cell membranes and alpha(1)-adrenoceptors present in rat cerebral cortex membranes.
European journal of pharmacology (2008-07-01)
Maura Floreani, Katia Varani, Luigi Quintieri, Pier Andrea Borea, Paola Dorigo
ABSTRACT
Recent in vitro studies, performed in rat aorta, mesenteric and intrapulmonary arteries, and human pulmonary artery, demonstrated that the beta-adrenoceptor ligand CGP-12177A (4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-1,3-dihydro-2H-benzimidazol-2-one) is also provided with antagonist or partial agonist properties at alpha(1)-adrenoceptors. These observations were supported by estimates of CGP-12177A binding affinity at alpha(1)-adrenoceptors, which have been always performed in rat cerebral cortex membranes, as a surrogate of vascular tissue. Since alpha(1D)-adrenoceptors are predominant in both rat aorta and mesenteric artery, in the present study, we measured, for the first time, the binding affinity of CGP-12177A at recombinant rat alpha(1D)-adrenoceptors expressed in BHK-21 cell membranes. CGP-12177A binding affinity was also determined in rat cerebral cortex membranes, where various alpha(1)-adrenoceptor subtypes are present. By means of [(3)H]prazosin binding competition experiments, we found that CGP-12177A bound to alpha(1D)-adrenoceptor-expressing BHK-21 cell membranes, with a binding affinity (pK(i)=5.39+/-0.27) almost identical to that measured in cerebral membranes (pK(i)=5.44+/-0.07), indicating that it is a non-subtype selective alpha(1)-adrenoceptor ligand. Moreover, CGP-12177A binding affinity was very close to its functional affinity evaluated in rat aorta in terms of antagonist potency against phenylephrine-induced contraction (pK(B)=5.65+/-0.07). In conclusion, our results demonstrate that, in order to evaluate CGP-12177A binding affinity at aorta and mesenteric artery alpha(1)-adrenoceptors, estimates in rat cerebral membranes are as reliable as those in recombinant rat alpha(1D)-adrenoceptors, since both values are very close to CGP-12177A functional affinities in isolated vessels.
Sigma-Aldrich
(±)-CGP-12177A, ≥98% (HPLC), solid
Pricing and availability is not currently available.