Merck
  • Retinal ganglion cell protection with geranylgeranylacetone, a heat shock protein inducer, in a rat glaucoma model.
Retinal ganglion cell protection with geranylgeranylacetone, a heat shock protein inducer, in a rat glaucoma model.
Investigative ophthalmology & visual science (2003-04-26)
Yoko Ishii, Jacky M K Kwong, Joseph Caprioli
ABSTRACT
To study the effects of geranylgeranylacetone (GGA) on the expression of inducible (HSP72) and constitutive (HSC70) heat shock proteins (HSPs) on retinal ganglion cells (RGCs) in a rat model of glaucoma. Adult Wistar rats were given intraperitoneal injections of GGA at 200 mg/kg daily. Western blot analysis and immunohistochemical staining for HSP72 and HSC70 were performed after 1, 3, and 7 days of treatment with GGA. After 7 days of GGA pretreatment, intraocular pressure (IOP) was elevated unilaterally by repeated trabecular argon laser photocoagulation 5 days after intracameral injection of india ink. After the first laser photocoagulation, GGA was administered twice a week. RGC survival was evaluated after 5 weeks of elevated IOP. Immunohistochemistry and TdT-mediated biotin-dUTP nick end labeling (TUNEL) were performed after 1 week of elevated IOP. Quercetin, an inhibitor of HSP expression, was also administered to a separate group. There was increased expression of HSP72 in RGCs at 3 and 7 days after administration of GGA, but HSC70 was unchanged. After 5 weeks of elevated IOP, there was a 27% +/- 6% loss of RGCs. The administration of GGA significantly reduced the loss of RGCs, lessened optic nerve damage, decreased the number of TUNEL-positive cells in the RGC layer, and increased HSP72. Quercetin abolished these protective effects. These results demonstrate that systemic administration of GGA protects RGCs from glaucomatous damage in a rat model and suggest a novel pathway for neuroprotection in patients with glaucoma.
Sigma-Aldrich
Geranylgeranylacetone
Pricing and availability is not currently available.