Merck
  • 2-thiouracil is a selective inhibitor of neuronal nitric oxide synthase antagonising tetrahydrobiopterin-dependent enzyme activation and dimerisation.
2-thiouracil is a selective inhibitor of neuronal nitric oxide synthase antagonising tetrahydrobiopterin-dependent enzyme activation and dimerisation.
FEBS letters (2000-11-30)
A Palumbo, M d'Ischia, F A Cioffi
ABSTRACT
2-thiouracil (TU), an established antithyroid drug and melanoma-seeker, was found to selectively inhibit neuronal nitric oxide synthase (nNOS) in a competitive manner (K(i)=20 microM), being inactive on the other NOS isoforms. The drug apparently interfered with the substrate- and tetrahydrobiopterin (BH(4))-binding to the enzyme. It caused a 60% inhibition of H(2)O(2) production in the absence of L-arginine and BH(4), and antagonised BH(4)-induced dimerisation of nNOS, but did not affect cytochrome c reduction. These results open new perspectives in the understanding of the antithyroid action of TU and provide a new lead structure for the development of selective nNOS inhibitors to elucidate the interdependence of the substrate and pteridine sites and to modulate pathologically aberrant NO formation.
Sigma-Aldrich
2-Thiouracil, ≥99%
Pricing and availability is not currently available.