Merck
  • MSX1-A Potential Marker for Uterus-Preserving Therapy of Endometrial Carcinomas.
MSX1-A Potential Marker for Uterus-Preserving Therapy of Endometrial Carcinomas.
International journal of molecular sciences (2020-07-08)
Simon Eppich, Christina Kuhn, Elisa Schmoeckel, Doris Mayr, Sven Mahner, Udo Jeschke, Julia Gallwas, Helene Hildegard Heidegger
ABSTRACT
Prognostic factors are of great interest in patients with endometrial cancer. One potential factor could be the protein MSX1, a transcription repressor, that has an inhibitory effect on the cell cycle. For this study, endometrioid endometrial carcinomas (n = 53), clear cell endometrial carcinomas (n = 6), endometrioid ovarian carcinomas (n = 19), and clear cell ovarian carcinomas (n = 11) were immunochemically stained for the protein MSX1 and evaluated using the immunoreactive score (IRS). A significant stronger expression of MSX1 was found in endometrioid endometrial carcinomas (p < 0.001), in grading 2 (moderate differentiation) (p = 0.001), and in tumor material of patients with no involvement of lymph nodes (p = 0.031). Correlations were found between MSX1 expression and the expression of β-Catenin, p21, p53, and the steroid receptors ERα, ERβ, PRα, and PRβ. A significant (p = 0.023) better survival for patients with an MSX1 expression in more than 10% of the tumor cells was observed for endometrioid endometrial carcinomas (21.3 years median survival (MSX1-positive) versus 17.3 years (MSX1-negative)). Although there is evidence that MSX1 expression correlates with improved long-term survival, further studies are necessary to evaluate if MSX1 can be used as a prognostic marker.
Sigma-Aldrich
Anti-MSX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Pricing and availability is not currently available.