Merck
  • Heartworm control in Grenada, West Indies: Results of a field study using imidacloprid 10% + moxidectin 2.5% and doxycycline for naturally-acquired Dirofilaria immitis infections.
Heartworm control in Grenada, West Indies: Results of a field study using imidacloprid 10% + moxidectin 2.5% and doxycycline for naturally-acquired Dirofilaria immitis infections.
Veterinary parasitology (2020-09-01)
T Paterson, C Fernandez, P J Burnett, L Lessey, T Hockley, R Hagen, C Coomansingh, B Sharma, R Chandrashekar, R Schaper
ABSTRACT
Canine heartworm disease (CHD) results from infection with Dirofilaria immitis and while it is of global concern, it is most prevalent in tropical climates where conditions support the parasite and vector life cycles. Melarsomine dihydrochloride is the sole treatment for CHD recommended by the American Heartworm Society. However, in cases where cost or access to melarsomine precludes treatment of an infected dog, therapeutic alternatives are warranted. This randomized, controlled field study evaluated the adulticidal efficacy of a combination therapeutic protocol using 10 % imidacloprid + 2.5 % moxidectin spot-on and a single 28-day course of doxycycline and compared with that of a 2-dose melarsomine dihydrochloride protocol. Of 37 naturally-infected domestic dogs with class 1, 2 or early class 3 CHD enrolled in the study, 30 were evaluated for a minimum of 12 months. Seven dogs were withdrawn due to canine ehrlichiosis, non-compliance, or wrongful inclusion. Dogs were randomly assigned to a control (CP, n = 15) or investigational (IVP, n = 15) treatment group. CP dogs received two injections of melarsomine dihydrochloride (2.5 mg/kg) 24 -hs apart and maintained on monthly ivermectin/pyrantel. IVP dogs were treated with oral doxycycline (10 mg/kg twice daily for 28 days) and topical 10 % imidacloprid + 2.5 % moxidectin once monthly for 9 months. Dogs were evaluated up to 18 months - monthly for the first 9 months, then every 3 months. Parasiticidal efficacy was based on antigen status using the IDEXX PetChek® 34 Heartworm-PF Antigen test. By month 18, antigen was not detected in any study dog except one from the IVP group. One other IVP dog was persistently antigenemic and treated with melarsomine at month 12 according to the initial study protocol. Mean antigen concentration (based on optical density) decreased more rapidly in the CP group and by month 15 was 0.11 for the IVP and 0.07 for CP groups, with equivalent median concentrations (0.04) in both groups. Conversion following heat-treatment of antigen-negative samples occurred frequently and at similar rates in both treatment groups. Based on the bias of diagnostic tests towards detection of female worms, we conclude that monthly application of 10 % imidacloprid + 2.5 % moxidectin for 9 months combined with a course of doxycycline twice daily for 28 days resulted in effective therapy against female adults in CHD. This therapeutic option may be particularly useful in cases where financial constraint or access to melarsomine precludes treatment of an infected individual. This study was supported by Bayer Animal Health.
Sigma-Aldrich
Moxidectin, ≥98% (HPLC)
Pricing and availability is not currently available.