Merck
  • Tumor-infiltrating lymphocyte secretion of IL-6 antagonizes tumor-derived TGF-beta 1 and restores the lymphokine-activated killing activity.
Tumor-infiltrating lymphocyte secretion of IL-6 antagonizes tumor-derived TGF-beta 1 and restores the lymphokine-activated killing activity.
Journal of immunology (Baltimore, Md. : 1950) (2004-01-22)
Ya-Wen Hsiao, Kuang-Wen Liao, Shao-Wen Hung, Rea-Min Chu
ABSTRACT
IL-6 is a multifunctional cytokine that regulates cell growth, differentiation, and cell survival. Many tumor cells produce TGF-beta1, which allows them to evade CTL-mediated immune responses. IL-6 antagonizes TGF-beta1 inhibition of CD3 cell activation. However, whether IL-6 restores NK activity, which also is suppressed by TGF-beta1, is not known. We used canine transmissible venereal tumor (CTVT), which produces TGF-beta1, as a model to determine whether IL-6 restores lymphokine-activated killer (LAK) activity. During the progression phase, CTVT cells stop expressing MHC molecules. During the regression phase, the number of surface MHC molecules increases dramatically on about one-third of tumor cells. Tumor cells that stop expressing MHC should be targeted by NK cells. In this study, we found that TGF-beta1 secreted by CTVT cells suppressed LAK cytotoxicity. Interestingly, tumor-infiltrating lymphocytes (TIL) isolated from regressing CTVT secrete high concentrations of IL-6 and antagonize the anti-LAK activity of tumor cell TGF-beta1. TIL also produce IL-6 during progression phase, but the concentration is too low to block the anti-LAK activity of TGF-beta1. There is probably a threshold concentration of IL-6 needed to reverse TGF-beta1-inhibited LAK activity. In addition, in the absence of TGF-beta1, IL-6 derived from TIL does not promote the activity of LAK. This new mechanism, in which TIL manufacture high concentrations of IL-6 to block tumor TGF-beta1 anti-LAK activity, has potential applications in cancer immunotherapy and tumor prognosis.
Sigma-Aldrich
Anti-Transforming Growth Factor-β, pan antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
Pricing and availability is not currently available.