Merck
  • Mitogen-activated protein kinase cascade is involved in endothelin-1-induced rat puerperal uterine contraction.
Mitogen-activated protein kinase cascade is involved in endothelin-1-induced rat puerperal uterine contraction.
Endocrinology (1999-02-02)
A Kimura, M Ohmichi, T Takeda, H Kurachi, H Ikegami, K Koike, K Masuhara, J Hayakawa, T Kanzaki, M Kobayashi, M Akabane, M Inoue, A Miyake, Y Murata
ABSTRACT
The regulation of mitogen-activated protein (MAP) kinase by endothelin-1 (ET-1) in cultured rat puerperal uterine myometrial cells was investigated. ET-1 caused the rapid stimulation of MAP kinase activity. ET-1-induced MAP kinase activation is neither extracellular Ca2+- nor intracellular Ca2+-dependent. ET-1 stimulation also led to an increase in phosphorylation of son-of-sevenless (SOS), and transfection of dominant negative SOS attenuated the ET-1-induced MAP kinase activity. Phorbol-12-myristate 13-acetate (PMA) also induced the MAP kinase activity, but pretreatment of the cultured cells with PMA, to down-regulate protein kinase C (PKC), did not abolish the activation of MAP kinase by ET-1. In addition, down-regulation of PKC had no effect on ET-1-induced SOS phosphorylation. Pertussis toxin, which inactivates Gi/Go proteins, blocked the ET-1-induced MAP kinase activation but not the PMA-induced MAP kinase activation. The results suggested that MAP kinase is acutely activated by ET-1 through a pertussis toxin-sensitive G protein and SOS, not through the PMA-sensitive PKC. In addition, although reverse-transcriptase PCR assays detected messenger RNA for both ET- 1 receptor subtypes in cultured rat puerperal uterine myometrial cells, ET-1-induced MAP kinase activity and uterine contraction were blocked by treatment with BQ485, an antagonist selective for an ET type A receptor (but not by BQ788, an ET type B receptor antagonist). Ritodrine, which is known to relax uterine muscle contraction, attenuated ET-1-induced MAP kinase activity. We further examined the role of MAP kinase pathway in uterine contraction using an inhibitor of MEK activity, PD098059. This inhibitor completely inhibited the ET-1-induced MAP kinase activation and partially, but significantly, inhibited the ET-1-induced uterine contraction. These results indicate that ET-1-induced MAP kinase signaling cascade may play an important role in the ET-1-induced uterine contraction.
Sigma-Aldrich
Ritodrine hydrochloride
Pricing and availability is not currently available.