Merck
  • Spin-frustration with two quasi-degenerated spin states of a copper(II) heptanuclear complex obtained from an amino acid ligand.
Spin-frustration with two quasi-degenerated spin states of a copper(II) heptanuclear complex obtained from an amino acid ligand.
Dalton transactions (Cambridge, England : 2003) (2020-05-28)
Catiúcia R M O Matos, Henrique C S Junior, Dayenny L D'Amato, Acácio S de Souza, Sérgio Pinheiro, Guilherme P Guedes, Glaucio B Ferreira, Odivaldo C Alves, Filipe B de Almeida, Flávio Garcia, Célia M Ronconi
ABSTRACT
The Cu(ii) heptanuclear complex (Cu7atac) was synthesised using the hydrated amino acid ligand 2-(5-amino-1H-1,2,4-triazol-3-yl)acetic acid (Hatac·H2O). Single crystal X-ray diffraction analysis revealed a μ3-hydroxo bridged Cu(ii) heptanuclear complex, consisting of two triangular subunits and one Cu(ii) ion as a bridge with the formula [Cu7(atac)6(μ3-OH)2(NO3)2(H2O)10](NO3)4. The magnetic behaviour of this discrete 0D complex shows strong antiferromagnetic couplings between Cu(ii) mediated by N,N bonding and an anti-anti modes of the carboxylate anion of the ligand atac-. The magnetic data were fitted considering a 3J model. To support the model used to fit the magnetic data of the Cu7atac complex, theoretical calculation methods (complete active space self-consistent field, CASSCF, density functional theory (DFT) using the UKS TPSS/Def2-TZVP//Def2-SVP level and periodic boundary conditions (PBC) using PBE/DZVP-MOLOPT-GTH) were performed to obtain the spin states, spin density map and J couplings. The theoretical results suggest that Cu7atac is a spin-frustrated complex in the ground state, in which the doublet spin state co-exists with the quartet spin state.
Sigma-Aldrich
Aminoguanidine bicarbonate, 97%
Pricing and availability is not currently available.