Merck
  • CD47-dependent molecular mechanisms of blood outgrowth endothelial cell attachment on cholesterol-modified polyurethane.
CD47-dependent molecular mechanisms of blood outgrowth endothelial cell attachment on cholesterol-modified polyurethane.
Biomaterials (2010-06-12)
Masako Ueda, Ivan S Alferiev, Stacey B Simons, Robert P Hebbel, Robert J Levy, Stanley J Stachelek
ABSTRACT
We previously showed that blood outgrowth endothelial cells (BOECs) had a high affinity for polyurethane (PU) covalently configured with cholesterol residues (PU-Chol). However, the molecular mechanisms responsible for this enhanced affinity were not determined. CD47, a multifunctional transmembrane glycoprotein involved in cellular attachment, can form a cholesterol-dependent complex with integrin alpha(v)beta(3) and heterotrimeric G proteins. We tested herein the hypothesis that CD47, and the other components of the multi-molecular complex, enhance the attachment of BOECs to PU-Chol. Immunoprecipitation studies, of human and ovine BOECs, demonstrated that CD47 associates with integrin alpha(v) and integrin beta(3) as well as G(alphai-2) protein. The three-fold increase in BOEC attachment to PU-Chol, compared to unmodified PU, was reversed with the addition of blocking antibodies specific for CD47 and integrin alpha(v) and integrin beta(3). Similar results were observed with the addition of methyl-beta-cyclodextrin (MbetaCD), a known disruptor of the CD47 complex as well as of the membrane cholesterol content, to seeded BOEC or PU-Chol films. Reducing CD47 expression, via lentivirus transduced shRNA, decreased BOEC binding to PU-Chol by 50% compared to control groups. These data are the first demonstration of a role for the CD47 cholesterol-dependent signaling complex in BOEC attachment onto synthetic surfaces.
Sigma-Aldrich
Anti-Integrin αV Antibody, clone LM142, ascites fluid, clone LM142, Chemicon®
Pricing and availability is not currently available.