Merck
  • Growth Mechanism of Alternating Defect Domains in Hexagonal WS2 via Inhomogeneous W-Precursor Accumulation.
Growth Mechanism of Alternating Defect Domains in Hexagonal WS2 via Inhomogeneous W-Precursor Accumulation.
Small (Weinheim an der Bergstrasse, Germany) (2020-10-01)
Gwang Hwi An, Seok Joon Yun, Young Hee Lee, Hyun Seok Lee
ABSTRACT
While a hexagonal WS2 monolayer, grown by chemical vapor deposition, exhibits distinctive patterns in photoluminescence mapping, segmented with alternating S-vacancy (SV) and W-vacancy (WV) domains in a single crystal, the formation mechanism for native alternating defect domains remains unresolved to date. Here, the formation mechanism of alternating defect domains in hexagonal WS2 via the precursor accumulation model is experimentally elucidated. A triangular WS2 seed is initially formed, followed by a hexagonal flake. Alternating W-rich (SV) and W-deficient (WV) domains are constructed in hexagonal WS2 flake, which is confirmed by confocal photoluminescence mapping and secondary ion mass spectroscopy. This is explained by the accumulation or scarcity of W-precursors at the edge of the WS2 flake. The W-precursors accumulate near the edges of the initial triangular WS2 seed over time, while they are deficient near the corners of the triangular WS2 , eventually forming WV domains in the truncated hexagonal domains. The heterogeneous accumulation becomes more prominent in the presence of H2 gas through desorption of the W-precursors.
Sigma-Aldrich
Ammonium metatungstate hydrate, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Pricing and availability is not currently available.