Merck
  • Microbial enrichment and multiplexed microfiltration for accelerated detection of Salmonella in spinach.
Microbial enrichment and multiplexed microfiltration for accelerated detection of Salmonella in spinach.
Biotechnology progress (2019-06-23)
Seockmo Ku, Eduardo Ximenes, Thomas Kreke, Kirk Foster, Justin L Couetil, Jessica Zuponcic, Xiaojun Zhao, Lori Hoagland, Amanda J Deering, Michael R Ladisch
ABSTRACT
To attain Salmonella detection thresholds in spinach suspensions using enrichment media requires at least 24 hr. Separation and concentration of selected microorganisms via microfiltration and microfugation reduce time for sample preparation, especially when working with large volumes of vegetable suspensions. This facilitates accelerated detection of Salmonella in spinach suspensions, and may contribute to effectively monitoring this pathogen before it reaches the consumer. We report a microfiltration-based protocol for accelerated sample preparation to concentrate and recover ≤1 colony forming unit (CFU) Salmonella/g pathogen-free spinach. Store-bought samples of spinach and a spinach plant subjected to two environmental conditions (temperature and light exposure) during its production were tested. The overall procedure involves extraction with buffer, a short enrichment step, prefiltration using a nylon filter, crossflow hollow fiber microfiltration, and retentate centrifugation to bring microbial cells to detection levels. Based on 1 CFU Salmonella/g frozen spinach, and a Poisson distribution statistical analyses with 99% probability, we calculated that 3 hr of incubation, when followed by microfiltration, is sufficient to reach the 2 log concentration required for Salmonella detection within 7 hr. Longer enrichment times (5 hr or more) is needed for concentrations lower than 1 CFU Salmonella/g of ready to eat spinach. The recovered microbial cells were identified and confirmed as Salmonella using both polymerase chain reaction (PCR) and plating methods. Different environmental conditions tested during production did not affect Salmonella viability; this demonstrated the broad adaptability of Salmonella and emphasized the need for methods that enable efficient monitoring of production for the presence of this pathogen.
Product Description
Millipore
Nylon Membrane Filter, 10.0 μm Pore Size, Millipore, filter diam. 90 mm, hydrophilic
Pricing and availability is not currently available.