Merck
  • Development of canine PD-1/PD-L1 specific monoclonal antibodies and amplification of canine T cell function.
Development of canine PD-1/PD-L1 specific monoclonal antibodies and amplification of canine T cell function.
PloS one (2020-07-03)
Jin Wook Choi, Sita S Withers, Hong Chang, Justin A Spanier, Victoria L De La Trinidad, Harmanpreet Panesar, Brian T Fife, Roger Sciammas, Ellen E Sparger, Peter F Moore, Michael S Kent, Robert B Rebhun, Stephen J McSorley
ABSTRACT
Interruption of the programmed death 1 (PD-1) / programmed death ligand 1 (PD-L1) pathway is an established and effective therapeutic strategy in human oncology and holds promise for veterinary oncology. We report the generation and characterization of monoclonal antibodies specific for canine PD-1 and PD-L1. Antibodies were initially assessed for their capacity to block the binding of recombinant canine PD-1 to recombinant canine PD-L1 and then ranked based on efficiency of binding as judged by flow cytometry. Selected antibodies were capable of detecting PD-1 and PD-L1 on canine tissues by flow cytometry and Western blot. Anti-PD-L1 worked for immunocytochemistry and anti-PD-1 worked for immunohistochemistry on formalin-fixed paraffin embedded canine tissues, suggesting the usage of this antibody with archived tissues. Additionally, anti-PD-L1 (JC071) revealed significantly increased PD-L1 expression on canine monocytes after stimulation with peptidoglycan or lipopolysaccharide. Together, these antibodies display specificity for the natural canine ligand using a variety of potential diagnostic applications. Importantly, multiple PD-L1-specific antibodies amplified IFN-γ production in a canine peripheral blood mononuclear cells (PBMC) concanavlin A (Con A) stimulation assay, demonstrating functional activity.
Sigma-Aldrich
Human PDCD1 / Programmed Cell Death Protein 1 ELISA Kit
Pricing and availability is not currently available.