Merck
  • Facile and Sensitive Near-Infrared Fluorescence Probe for the Detection of Endogenous Alkaline Phosphatase Activity In Vivo.
Facile and Sensitive Near-Infrared Fluorescence Probe for the Detection of Endogenous Alkaline Phosphatase Activity In Vivo.
Analytical chemistry (2017-05-19)
Song-Jiao Li, Chun-Yan Li, Yong-Fei Li, Junjie Fei, Ping Wu, Bin Yang, Juan Ou-Yang, Shi-Xin Nie
ABSTRACT
Alkaline phosphatase (ALP) is an essential enzyme and widely distributes in a variety of tissues. To date, various nanomaterial and small-molecule fluorescent probes for ALP have been constructed successfully, but the emission wavelengths of these probes are in the ultraviolet or visible range, which is not beneficial for bioimaging. Herein, a hemicyanine-based near-infrared (NIR) fluorescent probe named CyP is first synthesized and used to detect ALP activity. The characteristics of probe CyP are as follows: (1) The probe possesses a facile structure, which can be obtained by easy synthetic steps. (2) The fluorescence emission of the sensing system is at 738 nm belonging to NIR region, which is suitable for bioimaging in vivo. (3) The probe exhibits high sensitivity to ALP with 10-fold fluorescence enhancement and low detection limit (0.003 U/mL) can match the level of ALP in vivo. (4) The fluorescent change of the probe is attributed to the fact that ALP-catalyzed cleavage of the phosphate group in CyP induces the transformation of CyP (fluorescence off) into CyOH (fluorescence on), which is proved by HPLC, 31P NMR, MS, and DFT calculation. (5) The NIR fluorescent probe is applied for the detection of endogenous ALP activity in various biological samples such as cell, tissue, and living animal with satisfactory results.
Sigma-Aldrich
BioTracker CyP AP Live Cell Dye, Live cell imaging dye for alkaline phosphatase (AP) for cancer and stem cell applications.
Pricing and availability is not currently available.