Merck
  • Antiangiogenic therapy with Nintedanib affects hypoxia, angiogenesis and apoptosis in the ventral prostate of TRAMP animals.
Antiangiogenic therapy with Nintedanib affects hypoxia, angiogenesis and apoptosis in the ventral prostate of TRAMP animals.
Cell and tissue research (2019-09-02)
Raquel Frenedoso da Silva, Thais Petrochelli Banzato, Letícia Ferreira Alves, João Ernesto Carvalho, Rajesh Agarwal, Valéria Helena Alves Cagnon
ABSTRACT
The antiangiogenic therapy for prostate cancer with Nintedanib, a potent inhibitor of important growth factor receptors, has been proven to delay tumor progression and arrest tumor growth; thus, the aim herein is to evaluate Nintedanib effects on tumor cells, besides angiogenesis and apoptosis processes, metalloproteinases and hypoxia factor in an animal model. Nintedanib promoted growth inhibition and cell death in a dose-dependent manner, showing no tumor selectivity. Transgenic Adenocarcinoma of the Mouse Prostate (TRAMP) were treated with Nintedanib (10 mg/kg/day) in different stages of tumor development and the ventral prostate was examined for protein levels by means of immunohistochemistry and Western blotting and apoptosis evaluation. In vitro antiproliferative activity of Nintedanib was also assessed in nine human tumor cell lines. Early Nintedanib treatment has shown decreased levels of FGF-2, VEGFR-1, MMP-9 and HIF-1α and a significantly increased apoptosis of epithelial cells. Furthermore, late Nintedanib treatment decreased FGF-2, VEGFR-1 and FGFR-3 levels. Importantly, even after treatment discontinuation, treated animals displayed a significant decrease in VEGFR-1 as well as MMP-9. Although Nintedanib treatment in late stages of tumor growth has shown some good results, it is noteworthy that the drug presents the best tissue response when administered in the early stages of disease development. Nintedanib treatment has shown to be a promising approach for prostate cancer therapy, especially in the early stages of the disease, interfering in different carcinogenesis progression pathways.
Sigma-Aldrich
Nintedanib, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Nintedanib ethanesulphonate, ≥98% (HPLC)
Pricing and availability is not currently available.