Merck
  • Smac mimetics synergize with immune checkpoint inhibitors to promote tumour immunity against glioblastoma.
Smac mimetics synergize with immune checkpoint inhibitors to promote tumour immunity against glioblastoma.
Nature communications (2017-02-16)
Shawn T Beug, Caroline E Beauregard, Cristin Healy, Tarun Sanda, Martine St-Jean, Janelle Chabot, Danielle E Walker, Aditya Mohan, Nathalie Earl, Xueqing Lun, Donna L Senger, Stephen M Robbins, Peter Staeheli, Peter A Forsyth, Tommy Alain, Eric C LaCasse, Robert G Korneluk
ABSTRACT
Small-molecule inhibitor of apoptosis (IAP) antagonists, called Smac mimetic compounds (SMCs), sensitize tumours to TNF-α-induced killing while simultaneously blocking TNF-α growth-promoting activities. SMCs also regulate several immunomodulatory properties within immune cells. We report that SMCs synergize with innate immune stimulants and immune checkpoint inhibitor biologics to produce durable cures in mouse models of glioblastoma in which single agent therapy is ineffective. The complementation of activities between these classes of therapeutics is dependent on cytotoxic T-cell activity and is associated with a reduction in immunosuppressive T-cells. Notably, the synergistic effect is dependent on type I IFN and TNF-α signalling. Furthermore, our results implicate an important role for TNF-α-producing cytotoxic T-cells in mediating the anti-cancer effects of immune checkpoint inhibitors when combined with SMCs. Overall, this combinatorial approach could be highly effective in clinical application as it allows for cooperative and complimentary mechanisms in the immune cell-mediated death of cancer cells.
Sigma-Aldrich
AZD5582, ≥98% (HPLC)
Pricing and availability is not currently available.