Merck
  • pH-responsive high stability polymeric nanoparticles for targeted delivery of anticancer therapeutics.
pH-responsive high stability polymeric nanoparticles for targeted delivery of anticancer therapeutics.
Communications biology (2020-03-05)
L Palanikumar, Sumaya Al-Hosani, Mona Kalmouni, Vanessa P Nguyen, Liaqat Ali, Renu Pasricha, Francisco N Barrera, Mazin Magzoub
ABSTRACT
The practical application of nanoparticles (NPs) as chemotherapeutic drug delivery systems is often hampered by issues such as poor circulation stability and targeting inefficiency. Here, we have utilized a simple approach to prepare biocompatible and biodegradable pH-responsive hybrid NPs that overcome these issues. The NPs consist of a drug-loaded polylactic-co-glycolic acid (PLGA) core covalently 'wrapped' with a crosslinked bovine serum albumin (BSA) shell designed to minimize interactions with serum proteins and macrophages that inhibit target recognition. The shell is functionalized with the acidity-triggered rational membrane (ATRAM) peptide to facilitate internalization specifically into cancer cells within the acidic tumor microenvironment. Following uptake, the unique intracellular conditions of cancer cells degrade the NPs, thereby releasing the chemotherapeutic cargo. The drug-loaded NPs showed potent anticancer activity in vitro and in vivo while exhibiting no toxicity to healthy tissue. Our results demonstrate that the ATRAM-BSA-PLGA NPs are a promising targeted cancer drug delivery platform.
Sigma-Aldrich
(3-Carboxypropyl)triphenylphosphonium bromide, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Pricing and availability is not currently available.