Merck
  • Blood fruit (Haematocarpus validus (Miers.) Bakh. f. ex Forman): a novel source of natural food colourant.
Blood fruit (Haematocarpus validus (Miers.) Bakh. f. ex Forman): a novel source of natural food colourant.
Journal of food science and technology (2020-01-25)
Pooja Bohra, Ajit Arun Waman, Tapas Kumar Roy, K S Shivashankara
ABSTRACT
Plant based pigments have widely been used by human beings since ancient times. These pigments, being natural in origin, have better acceptability amongst the consumers and offer diversified applications worldwide. For identification of novel pigment sources, lesser known species need to be studied systematically. In the present report, a tropical fruit species viz. blood fruit was studied for its physicochemical parameters and anthocyanin content. Fruit pulp had total soluble solids of 17 °B with acidic pH (3.01) and total phenolic content of 13.44 mg GAE/100 g of fresh pulp. Pulp of fully ripe fruits contained 8.76 mg/g of total anthocyanins. Pelargonidin was the dominant anthocyanin, followed by Cyanidin, Peonidin and Petunidin. Considering these qualities, the species could be a potential candidate for food and dye industries.
Sigma-Aldrich
Cyanidin chloride, ≥95% (HPLC)
Pricing and availability is not currently available.