Merck
  • Is tranexamic acid toxic to articular cartilage when administered topically? What is the safe dose?
Is tranexamic acid toxic to articular cartilage when administered topically? What is the safe dose?
The bone & joint journal (2018-03-29)
J D Parker, K S Lim, D C Kieser, T B F Woodfield, G J Hooper
ABSTRACT
The intra-articular administration of tranexamic acid (TXA) has been shown to be effective in reducing blood loss in unicompartmental knee arthroplasty and anterior cruciate reconstruction. The effects on human articular cartilage, however, remains unknown. Our aim, in this study, was to investigate any detrimental effect of TXA on chondrocytes, and to establish if there was a safe dose for its use in clinical practice. The hypothesis was that TXA would cause a dose-dependent damage to human articular cartilage. The cellular morphology, adhesion, metabolic activity, and viability of human chondrocytes when increasing the concentration (0 mg/ml to 40 mg/ml) and length of exposure to TXA (0 to 12 hours) were analyzed in a 2D model. This was then repeated, excluding cellular adhesion, in a 3D model and confirmed in viable samples of articular cartilage. Increasing concentrations above 20 mg/ml resulted in atypical morphology, reduced cellular adhesion and metabolic activity associated with increased chondrocyte death. However, the cell matrix was not affected by the concentration of TXA or the length of exposure, and offered cellular protection for concentrations below 20 mg/ml. These results show that when in vitro chondrocytes are exposed to higher concentrations of TXA, such as that expected following recommended intra-articular administration, cytotoxicity is observed. This effect is dose-dependent, such that a tissue concentration of 10 mg/ml to 20 mg/ml could be expected to be safe. Cite this article: Bone Joint J 2018;100-B:404-12.
Sigma-Aldrich
Ruthenium photoinitiator kit
Pricing and availability is not currently available.