Merck
  • Band Engineering and Majority Carrier Switching in Isostructural Donor-Acceptor Complexes DPTTA-F X TCNQ Crystals (X = 1, 2, 4).
Band Engineering and Majority Carrier Switching in Isostructural Donor-Acceptor Complexes DPTTA-F X TCNQ Crystals (X = 1, 2, 4).
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2020-02-12)
Yingying Liang, Yunke Qin, Jie Chen, Weilong Xing, Ye Zou, Yimeng Sun, Wei Xu, Daoben Zhu
ABSTRACT
Three isostructural donor-acceptor complexes DPTTA-F X TCNQ (X = 1, 2, 4) are investigated experimentally and theoretically. By tuning the number of F atoms in the acceptor molecules, the resulting complexes display a continuous down shift of the valence band maximum, conducting band minimum, and optical bandgap. The majority carriers convert from hole (DPTTA-F1TCNQ), balanced hole, and electron (DPTTA-F2TCNQ) to electron (DPTTA-F4TCNQ). This result shows that band engineering can be realized easily in the donor-acceptor complex systems by tuning the electron affinity of the acceptor. The bandgaps of these three complexes vary from 0.31 to 0.41 eV; this narrow bandgap feature is crucial for achieving high thermoelectric performance and the unintentional doping in DPTTA-F4TCNQ leads to the effective suppression of the bipolar cancelling effect on the Seebeck coefficient and the highest power factor.
Sigma-Aldrich
2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 97%
Pricing and availability is not currently available.