Merck
  • Co-occurrence of histone H3 K27M and BRAF V600E mutations in paediatric midline grade I ganglioglioma.
Co-occurrence of histone H3 K27M and BRAF V600E mutations in paediatric midline grade I ganglioglioma.
Brain pathology (Zurich, Switzerland) (2016-12-17)
Mélanie Pagès, Kevin Beccaria, Nathalie Boddaert, Raphaël Saffroy, Aurore Besnard, David Castel, Frédéric Fina, Doriane Barets, Emilie Barret, Ludovic Lacroix, Franck Bielle, Felipe Andreiuolo, Arnault Tauziède-Espariat, Dominique Figarella-Branger, Stéphanie Puget, Jacques Grill, Fabrice Chrétien, Pascale Varlet
ABSTRACT
Ganglioglioma (GG) is a grade I tumor characterized by alterations in the MAPK pathway, including BRAF V600E mutation. Recently, diffuse midline glioma with an H3 K27M mutation was added to the WHO 2016 classification as a new grade IV entity. As co-occurrence of H3 K27M and BRAF V600E mutations has been reported in midline tumors and anaplastic GG, we searched for BRAF V600E and H3 K27M mutations in a series of 54 paediatric midline grade I GG (midline GG) to determine the frequency of double mutations and its relevance for prognosis. Twenty-seven patients (50%) possessed the BRAF V600E mutation. The frequency of the co-occurrence of H3F3A/BRAF mutations at diagnosis was 9.3%. No H3 K27M mutation was detected in the absence of the BRAF V600E mutation. Double-immunostaining revealed that BRAF V600E and H3 K27M mutant proteins were present in both the glial and neuronal components. Immunopositivity for the BRAF V600E mutant protein correlated with BRAF mutation status as detected by massARRAY or digital droplet PCR. The median follow-up of patients with double mutation was 4 years. One patient died of progressive disease 8 years after diagnosis, whereas the four other patients were all alive with stable disease at the last clinical follow-up (at 9 months, 1 year and 7 years) without adjuvant therapy. We demonstrate in this first series of midline GGs that the H3 K27M mutation can occur in association with the BRAF V600E mutation in grade I glioneuronal tumors. Despite the presence of H3 K27M mutations, these cases should not be graded and treated as grade IV tumors because they have a better spontaneous outcome than classic diffuse midline H3 K27M-mutant glioma. These data suggest that H3 K27M cannot be considered a specific hallmark of grade IV diffuse gliomas and highlight the importance of integrated histomolecular diagnosis in paediatric brain tumors.
Sigma-Aldrich
Anti-Histone H3.3 Antibody, K27M mutant, from rabbit, purified by affinity chromatography
Pricing and availability is not currently available.