Merck
  • Expression of proteins involved in DNA damage response in familial and sporadic breast cancer patients.
Expression of proteins involved in DNA damage response in familial and sporadic breast cancer patients.
International journal of cancer (2015-07-25)
Giulia Partipilo, Giovanni Simone, Anna Scattone, Emanuela Scarpi, Amalia Azzariti, Anita Mangia
ABSTRACT
Understanding the expression of proteins involved in DNA damage response could improve knowledge of the pathways that contribute to familial and sporadic breast cancer (BC). We aimed to assess the different roles of BRCA1, poly(ADP-ribose) polymerase-1 (PARP1), BRCT-repeat inhibitor of hTERT expression (BRIT1) and novel SWItch 5 (SWI5) expression in 130 sporadic and 73 familial BC samples, by immunohistochemistry. In the sporadic group, negative nuclear BRCA1 (nBRCA1) expression was associated with positive PgR (p = 0.037). Negative association was found between nBRCA1 expression and HER2 (p = 0.001). In the familial group, nBRCA1 expression was associated with ER (p = 0.002). Reduced nBRCA1 expression was associated with higher histological grade and positive Ki67 both in sporadic (p = 0.0010, p = 0.047) and familial groups (p < 0.001, p = 0.001). Nuclear PARP1 (nPARP1) expression was associated with histological grade (p = 0.035) and positive PgR (p = 0.047) in sporadic cases. High cytoplasmic and low nuclear BRIT1 (cBRIT1 and nBRIT1) expression were associated with high histological grade in the familial group (p = 0.013, p = 0.025). Various statistical associations between the protein expressions were observed in the sporadic group, while in familial group only few associations were found. Univariate analyses showed that nPARP1 expression is able to discriminate between sporadic and familial tumors (OR 2.80, p = 0.002). Multivariate analyses proved that its overexpression is an independent factor associated with a high risk of sporadic tumor (OR 2.96, p = 0.017). Our findings indicate that nPARP1 expression is an independent factor for sporadic BCs and PARP1 inhibitors could be a promising therapy for different phenotypes.
Sigma-Aldrich
Anti-BRCA1 Antibody, clone MS110, clone MS110, from mouse
Pricing and availability is not currently available.