Merck
  • Oral Administration of Caffeine Exacerbates Cisplatin-Induced Hearing Loss.
Oral Administration of Caffeine Exacerbates Cisplatin-Induced Hearing Loss.
Scientific reports (2019-07-04)
Sandeep Sheth, Kelly Sheehan, Asmita Dhukhwa, Raheem F H Al Aameri, Chaitanya Mamillapalli, Debashree Mukherjea, Leonard P Rybak, Vickram Ramkumar
ABSTRACT
Adenosine A1 receptors (A1AR) are well characterized for their role in cytoprotection. Previous studies have demonstrated the presence of these receptors in the cochlea where their activation were shown to suppress cisplatin-induced inflammatory response and the resulting ototoxicity. Inhibition of A1AR by caffeine, a widely consumed psychoactive substance, could antagonize the endogenous protective role of these receptors in cochlea and potentiate cisplatin-induced hearing loss. This hypothesis was tested in a rat model of cisplatin ototoxicity following oral administration of caffeine. We report here that single-dose administration of caffeine exacerbates cisplatin-induced hearing loss without increasing the damage to outer hair cells (OHCs), but increased synaptopathy and inflammation in the cochlea. These effects of caffeine were mediated by its blockade of A1AR, as co-administration of R-PIA, an A1AR agonist, reversed the detrimental actions of caffeine and cisplatin on hearing loss. Multiple doses of caffeine exacerbated cisplatin ototoxicity which was associated with damage to OHCs and cochlear synaptopathy. These findings highlight a possible drug-drug interaction between caffeine and cisplatin for ototoxicity and suggest that caffeine consumption should be cautioned in cancer patients treated with a chemotherapeutic regimen containing cisplatin.
Sigma-Aldrich
Anti-Neurofilament H Antibody, clone NE14, Alexa Fluor 488 Conjugated, clone NE14, from mouse, ALEXA FLUOR 488
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Glutamate Receptor 2 Antibody, extracellular, clone 6C4, clone 6C4, Chemicon®, from mouse
Pricing and availability is not currently available.