Merck
  • Evaluation of the volatile composition, toxicological and antioxidant potentials of the essential oils and teas of commercial Chilean boldo samples.
Evaluation of the volatile composition, toxicological and antioxidant potentials of the essential oils and teas of commercial Chilean boldo samples.
Food research international (Ottawa, Ont.) (2019-08-31)
Wanderson Fernando Mello de Souza, Xavier Maia Mariano, Júlia Lima Isnard, Gisele Santos de Souza, Ana Luísa de Souza Gomes, Rutheneia José Tavares de Carvalho, Cristiane Barbosa Rocha, César Luis Siqueira Junior, Ricardo Felipe Alves Moreira
ABSTRACT
Chilean boldo (Peumus boldus Molina) is the boldo species most consumed around the world. Digestive and hepatobiliary disorders represent the main targets of its action. This work aims to characterize the volatile chemical composition, toxicological, and antioxidant potentials of the essential oils and teas of commercial samples of Chilean boldo packed on sachets [Group 1 (G1): five samples] or in plastic bags [Group 2 (G2): five samples]. Fifty-three compounds have been identified in the essential oils of commercial samples of Chilean boldo from Brazil, while only twelve compounds have been found in the volatile fraction of their infusions. Terpineol, 1,8-cineole, and p-cymene are the major compounds of essential oils. Terpineol is also the major compound of the volatile fraction of teas, followed by limonene dioxide. The presence in all samples of the chemical markers p-cymene, 1,8-cineole, ascaridole, and boldine suggests that they are genuine. The teas offer a better antioxidant capacity than essential oils, thereby indicating that antioxidant activity is concentrated in the non-volatile fraction of these herbs. All LD50 values estimated for the essential oils are below 200 ppm, thus indicating that the oils are highly cytotoxic. G1 and G2 appear to be very similar with respect to all the parameters analyzed. This similarity may indicate a single source for these products.
Sigma-Aldrich
Methyl eugenol, ≥98%, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
2-Undecanone, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Decanal, ≥98% (GC), liquid
Pricing and availability is not currently available.