Merck
  • Identification of AKN-032, a novel 2-aminopyrazine tyrosine kinase inhibitor, with significant preclinical activity in acute myeloid leukemia.
Identification of AKN-032, a novel 2-aminopyrazine tyrosine kinase inhibitor, with significant preclinical activity in acute myeloid leukemia.
Biochemical pharmacology (2010-08-14)
Anna Eriksson, Martin Höglund, Elin Lindhagen, Anna Aleskog, Sadia Bashir Hassan, Carina Ekholm, Karin Fhölenhag, Annika Jenmalm Jensen, Agneta Löthgren, Martin Scobie, Rolf Larsson, Vendela Parrow
ABSTRACT
Aberrant signal transduction by mutant or overexpressed protein kinases has emerged as a promising target for treatment of acute myeloid leukemia (AML). We here present a novel low molecular weight kinase inhibitor, AKN-032, targeting the FMS-like tyrosine kinase 3 (FLT3) and discovered in a new type of screening funnel combining the target therapy approach with sequential cellular screens. AKN-032 was identified among 150 selected hits from three different high throughput kinase screens. Further characterization showed inhibitory activity on FLT3 enzyme with an IC(50) of 70 nM. Western blot analysis revealed reduced autophosphorylation of the FLT3-receptor in AML cell line MV4-11 cells after exposure to AKN-032. Flow cytometry disclosed cytotoxic activity against MV4-11, but not against non-malignant 3T3-L1 fibroblast cells. Using a fluorometric microculture cytotoxicity assay, AKN-032 was tested against 15 cell lines and displayed a potent cytotoxic activity in AML cell lines MV4-11 (IC(50)=0.4 μM) and Kasumi-1 (IC(50)=2.3 μM). AKN-032 was also highly cytotoxic in tumor cells from AML patients in vitro. Furthermore, AKN-032 demonstrated significant antileukemic effect in a relatively resistant in vivo hollow fiber mouse model. No major toxicity was observed in the animals. In conclusion, AKN-032 is a promising new kinase inhibitor with significant in vivo and in vitro activity in AML. Results from the hollow fiber mouse assay suggest a favorable toxicity profile. Future studies will focus on pharmacokinetic properties, toxicity as well as further clarifying the mechanisms of action of AKN-032 in AML.
Sigma-Aldrich
Aminopyrazine, 98%
Pricing and availability is not currently available.