Merck
  • Purification, characterization, and substrate specificity of a novel highly glucose-tolerant beta-glucosidase from Aspergillus oryzae.
Purification, characterization, and substrate specificity of a novel highly glucose-tolerant beta-glucosidase from Aspergillus oryzae.
Applied and environmental microbiology (1998-10-06)
C Riou, J M Salmon, M J Vallier, Z Günata, P Barre
ABSTRACT
Aspergillus oryzae was found to secrete two distinct beta-glucosidases when it was grown in liquid culture on various substrates. The major form had a molecular mass of 130 kDa and was highly inhibited by glucose. The minor form, which was induced most effectively on quercetin (3,3',4',5,7-pentahydroxyflavone)-rich medium, represented no more than 18% of total beta-glucosidase activity but exhibited a high tolerance to glucose inhibition. This highly glucose-tolerant beta-glucosidase (designated HGT-BG) was purified to homogeneity by ammonium sulfate precipitation, gel filtration, and anion-exchange chromatography. HGT-BG is a monomeric protein with an apparent molecular mass of 43 kDa and a pI of 4.2 as determined by sodium dodecyl sulfate-polyacrylamide gel electrophoresis and isoelectric focusing polyacrylamide gel electrophoresis, respectively. Using p-nitrophenyl-beta-D-glucoside as the substrate, we found that the enzyme was optimally active at 50 degreesC and pH 5.0 and had a specific activity of 1,066 micromol min-1 mg of protein-1 and a Km of 0.55 mM under these conditions. The enzyme is particularly resistant to inhibition by glucose (Ki, 1. 36 M) or glucono-delta-lactone (Ki, 12.5 mM), another powerful beta-glucosidase inhibitor present in wine. A comparison of the enzyme activities on various glycosidic substrates indicated that HGT-BG is a broad-specificity type of fungal beta-glucosidase. It exhibits exoglucanase activity and hydrolyzes (1-->3)- and (1-->6)-beta-glucosidic linkages most effectively. This enzyme was able to release flavor compounds, such as geraniol, nerol, and linalol, from the corresponding monoterpenyl-beta-D-glucosides in a grape must (pH 2.9, 90 g of glucose liter-1). Other flavor precursors (benzyl- and 2-phenylethyl-beta-D-glucosides) and prunin (4',5,7-trihydroxyflavanone-7-glucoside), which contribute to the bitterness of citrus juices, are also substrates of the enzyme. Thus, this novel beta-glucosidase is of great potential interest in wine and fruit juice processing because it releases aromatic compounds from flavorless glucosidic precursors.
Sigma-Aldrich
p-Nitrophenyl-β-D-glucopyranoside, Chromogenic substrate for β-glucosidase.
Pricing and availability is not currently available.