Merck
  • Anti-Neurofascin-155 IgG4 antibodies prevent paranodal complex formation in vivo.
Anti-Neurofascin-155 IgG4 antibodies prevent paranodal complex formation in vivo.
The Journal of clinical investigation (2019-03-15)
Constance Manso, Luis Querol, Cinta Lleixà, Mallory Poncelet, Mourad Mekaouche, Jean-Michel Vallat, Isabel Illa, Jérôme J Devaux
ABSTRACT
Neurofascin-155 (Nfasc155) is an essential glial cell adhesion molecule expressed in paranodal septate-like junctions of peripheral and central myelinated axons. The genetic deletion of Nfasc155 results in the loss of septate-like junctions and in conduction slowing. In humans, IgG4 antibodies against Nfasc155 are implicated in the pathogenesis of chronic inflammatory demyelinating polyneuropathy (CIDP). These antibodies are associated with an aggressive onset, a refractoriness to intravenous immunoglobulin, and tremor of possible cerebellar origin. Here, we examined the pathogenic effects of patient-derived anti-Nfasc155 IgG4. These antibodies did not inhibit the ability of Nfasc155 to complex with its axonal partners contactin-1/CASPR1 or induce target internalization. Passive transfer experiments revealed that IgG4 antibodies target Nfasc155 on Schwann cell surface, and diminished Nfasc155 protein levels and prevented paranodal complex formation in neonatal animals. In adult animals, chronic intrathecal infusions of antibodies also induced the loss of Nfasc155 and of paranodal specialization and resulted in conduction alterations in motor nerves. These results indicate that anti-Nfasc155 IgG4 perturb conduction in absence of demyelination, validating the existence of paranodopathy. These results also shed light on the mechanisms regulating protein insertion at paranodes.
Sigma-Aldrich
Anti-Human IgG3−FITC antibody, Mouse monoclonal, clone HP-6050, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-β-Catenin Antibody, clone 5H10, clone 5H10, Chemicon®, from mouse
Pricing and availability is not currently available.