Merck
  • Molecular cloning and expression of multiple isoforms of human prostaglandin E receptor EP3 subtype generated by alternative messenger RNA splicing: multiple second messenger systems and tissue-specific distributions.
Molecular cloning and expression of multiple isoforms of human prostaglandin E receptor EP3 subtype generated by alternative messenger RNA splicing: multiple second messenger systems and tissue-specific distributions.
Molecular pharmacology (1995-11-01)
M Kotani, I Tanaka, Y Ogawa, T Usui, K Mori, A Ichikawa, S Narumiya, T Yoshimi, K Nakao
ABSTRACT
Five distinct cDNA clones encoding four different isoforms of human prostaglandin (PG) E receptor EP3 subtype were isolated from a human kidney cDNA library. Two cDNA clones differed only in their 3'-untranslated regions. The four isoforms, tentatively named EP3-I, EP3-II, EP3-III, and EP3-IV, which were generated by alternative mRNA splicing, had identical amino acid sequences except for their different carboxyl-terminal tails. Transfection experiments revealed that all the four isoforms show high binding affinities to PGE2, PGE1, and M&B28767, an EP3-specific agonist, whereas their downstream signaling pathways are divergent. M&B28767 increased cAMP concentrations in cells expressing EP3-II and EP3-IV, whereas it inhibited forskolin-induced cAMP accumulations in cells expressing all EP3 isoforms. M&B28767 also stimulated phosphoinositide turnover in cells expressing EP3-I and EP3-II. Northern blot analysis revealed that the EP3 gene is expressed in a wide variety of human tissues. The human EP3 mRNA was present most abundantly in the kidney, pancreas, and uterus. A substantial expression was also detected in the heart, liver, skeletal muscle, small intestine, colon, prostate, ovary, and testis. Furthermore, reverse transcription-polymerase chain reaction analysis demonstrated tissue-specific expressions of the five different EP3 mRNA species. The present study suggests the presence of the multiple systems of PGE2/EP3 isoforms and leads to the better understanding of its physiological and pathophysiological implications in humans.
Sigma-Aldrich
ChemiSCREEN EP3 Prostanoid Receptor Membrane Preparation, Human EP3 GPCR membrane preparation for Radioligand binding Assays & GTPgammaS binding.
Pricing and availability is not currently available.