Merck
The metabolism of o-fluoroaniline by rats, rabbits and marmosets.
Xenobiotica; the fate of foreign compounds in biological systems (1986-06-01)
C V Eadsforth, P C Coveney, D H Hutson, C J Logan, A J Samuel
ABSTRACT
o-Fluoroaniline is rapidly metabolized and excreted in rats, rabbits and marmosets. Following a single oral dose of 14C-fluoroaniline of about 20 mg/kg, more than 80% of the dose is excreted in 0-24 h, the urine being the major route of excretion for all three species. For all three species, 4-amino-3-fluorophenyl is a major metabolite, conjugated at oxygen with either sulphate or glucuronic acid. 4-Acetylamino-3-fluorophenyl sulphate or glucuronide are also significant metabolites. An h.p.l.c. method with electrochemical detection was developed for monitoring exposure of plant workers to o-fluoroaniline, based on 4-amino-3-fluorophenyl sulphate.
Sigma-Aldrich
4-Fluoroaniline, 99%
Pricing and availability is not currently available.