Merck
  • New Treatment of Medullary and Papillary Human Thyroid Cancer: Biological Effects of Hyaluronic Acid Hydrogel Loaded With Quercetin Alone or in Combination to an Inhibitor of Aurora Kinase.
New Treatment of Medullary and Papillary Human Thyroid Cancer: Biological Effects of Hyaluronic Acid Hydrogel Loaded With Quercetin Alone or in Combination to an Inhibitor of Aurora Kinase.
Journal of cellular physiology (2015-12-15)
Vincenzo Quagliariello, Emilia Armenia, Caterina Aurilio, Francesco Rosso, Ottavia Clemente, Gabriele de Sena, Manlio Barbarisi, Alfonso Barbarisi
ABSTRACT
The aim of this paper is based on the use of a hyaluronic acid hydrogel of Quercetin tested alone and in combination to an inhibitor of Aurora Kinase type A and B (SNS-314) on human medullary and papillary thyroid cancer cells. Biological investigations were focused on the cellular uptake of the hydrogel, cell viability, antioxidant, and cytokines secretion studies. Quercetin delivered from hydrogel show a time and CD44 dependent interaction with both cell lines with significant anti-inflammatory effects. Combination of Quercetin and SNS-314 leads to a synergistic cytotoxic effect on medullary TT and papillary BCPAP cell lines with a significant reduction of the IC50 value. These results, highlights the importance of synergistic effect of the hyaluronic acid hydrogel of Quercetin with SNS-314 in the regulation of human thyroid cancer cell proliferation and emphasize the anti-tumor activity of these molecules. J. Cell. Physiol. 231: 1784-1795, 2016. © 2015 Wiley Periodicals, Inc.
Sigma-Aldrich
SNS-314 mesylate, ≥98% (HPLC)
Pricing and availability is not currently available.