Merck
  • Role of stearoyl-CoA desaturase-1 in skin integrity and whole body energy balance.
Role of stearoyl-CoA desaturase-1 in skin integrity and whole body energy balance.
The Journal of biological chemistry (2013-12-21)
Harini Sampath, James M Ntambi
ABSTRACT
The skin is the single largest organ in humans, serving as a major barrier to infection, water loss, and abrasion. The functional diversity of skin requires the synthesis of large amounts of lipids, such as triglycerides, wax esters, squalene, ceramides, free cholesterol, free fatty acids, and cholesterol and retinyl esters. Some of these lipids are used as cell membrane components, signaling molecules, and a source of energy. An important class of lipid metabolism enzymes expressed in skin is the Δ(9)-desaturases, which catalyze the synthesis in Δ(9)-monounsaturated lipids, primarily oleoyl-CoA (18:1n-9) and palmitoyl-CoA (16:1n-7), the major monounsaturated fatty acids in cutaneous lipids. Mice with a deletion of the Δ(9)-desaturase-1 isoform (SCD1) either globally (Scd1(-/-)) or specifically in the skin (skin-specific Scd1-knockout; SKO) present with marked changes in cutaneous lipids and skin integrity. Interestingly, these mice also exhibit increased whole body energy expenditure, protection against diet-induced adiposity, hepatic steatosis, and glucose intolerance. The increased energy expenditure in skin-specific Scd1-knockout (SKO) mice is a surprising phenotype, as it links cutaneous lipid homeostasis with whole body energy balance. This minireview summarizes the role of skin SCD1 in regulating skin integrity and whole body energy homeostasis and offers a discussion of potential pathways that may connect these seemingly disparate phenotypes.
Product Description
Avanti
18:1 (n9) Coenzyme A, Avanti Research - A Croda Brand 870719P, powder
Pricing and availability is not currently available.
Avanti
16:1 (n7) Coenzyme A, Avanti Research - A Croda Brand 870743P, powder
Pricing and availability is not currently available.