Merck
  • Azospirillum brasilense Increases CO2 Fixation on Microalgae Scenedesmus obliquus, Chlorella vulgaris, and Chlamydomonas reinhardtii Cultured on High CO2 Concentrations.
Azospirillum brasilense Increases CO2 Fixation on Microalgae Scenedesmus obliquus, Chlorella vulgaris, and Chlamydomonas reinhardtii Cultured on High CO2 Concentrations.
Microbial ecology (2018-01-13)
Francisco J Choix, Cecilia Guadalupe López-Cisneros, Hugo Oscar Méndez-Acosta
ABSTRACT
Mutualism interactions of microalgae with other microorganisms are widely used in several biotechnological processes since symbiotic interaction improves biotechnological capabilities of the microorganisms involved. The interaction of the bacterium Azospirillum brasilense was assessed with three microalgae genus, Scenedesmus, Chlorella, and Chlamydomonas, during CO2 fixation under high CO2 concentrations. The results in this study have demonstrated that A. brasilense maintained a mutualistic interaction with the three microalgae assessed, supported by the metabolic exchange of indole-3-acetic acid (IAA) and tryptophan (Trp), respectively. Besides, CO2 fixation increased, as well as growth and cell compound accumulation, mainly carbohydrates, in each microalgae evaluated, interacting with the bacterium. Overall, these results propose the mutualism interaction of A. brasilense with microalgae for improving biotechnological processes based on microalgae as CO2 capture and their bio-refinery capacity.
Sigma-Aldrich
Alginic acid sodium salt, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Pricing and availability is not currently available.
Millipore
Nutrient Broth, Liquid medium used for the cultivation of a wide variety of microorganism, NutriSelect® Plus, suitable for microbiology
Pricing and availability is not currently available.