Merck
  • Focused Ultrasound Enabled Trans-Blood Brain Barrier Delivery of Gold Nanoclusters: Effect of Surface Charges and Quantification Using Positron Emission Tomography.
Focused Ultrasound Enabled Trans-Blood Brain Barrier Delivery of Gold Nanoclusters: Effect of Surface Charges and Quantification Using Positron Emission Tomography.
Small (Weinheim an der Bergstrasse, Germany) (2018-07-03)
Deborah Sultan, Dezhuang Ye, Gyu Seong Heo, Xiaohui Zhang, Hannah Luehmann, Yimei Yue, Lisa Detering, Sergey Komarov, Sara Taylor, Yuan-Chuan Tai, Joshua B Rubin, Hong Chen, Yongjian Liu
ABSTRACT
Focused ultrasound (FUS) technology is reported to enhance the delivery of 64 Cu-integrated ultrasmall gold nanoclusters (64 Cu-AuNCs) across the blood-brain barrier (BBB) as measured by positron emission tomography (PET). To better define the optimal physical properties for brain delivery, 64 Cu-AuNCs with different surface charges are synthesized and characterized. In vivo biodistribution studies are performed to compare the individual organ uptake of each type of 64 Cu-AuNCs. Quantitative PET imaging post-FUS treatment shows site-targeted brain penetration, retention, and diffusion of the negative, neutral, and positive 64 Cu-AuNCs. Autoradiography is performed to compare the intrabrain distribution of these nanoclusters. PET Imaging demonstrates the effective BBB opening and successful delivery of 64 Cu-AuNCs into the brain. Of the three 64 Cu-AuNCs investigated, the neutrally charged nanostructure performs the best and is the candidate platform for future theranostic applications in neuro-oncology.
Sigma-Aldrich
N-Boc-ethylenediamine, ≥98.0% (NT)
Pricing and availability is not currently available.