Merck
  • Use of pharmacokinetic-pharmacodynamic analyses to optimize therapy with the systemic antifungal micafungin for invasive candidiasis or candidemia.
Use of pharmacokinetic-pharmacodynamic analyses to optimize therapy with the systemic antifungal micafungin for invasive candidiasis or candidemia.
Antimicrobial agents and chemotherapy (2011-02-09)
David Andes, Paul G Ambrose, Jeffrey P Hammel, Scott A Van Wart, Varsha Iyer, Daniel K Reynolds, Donald N Buell, Laura L Kovanda, Sujata M Bhavnani
ABSTRACT
Echinocandins have become a first-line therapy for invasive candidiasis (IC). Using phase 3 trial data for patients with IC, pharmacokinetic-pharmacodynamic (PK-PD) relationships for efficacy for micafungin were examined. Micafungin exposures were estimated using a population pharmacokinetic model, and univariable and multivariable logistic regressions were used to identify factors associated with outcome, including the micafungin area under the concentration-time curve (AUC)/MIC ratio. Monte Carlo simulation was used to evaluate the probability of achieving AUC/MIC ratios associated with efficacy. Mycological and clinical success rates for evaluable cases were 89.4 and 90.9, respectively. MIC50s and MIC90s for Candida species inhibition were 0.008 and 0.5 mg/liter, respectively. The median AUC/MIC ratio was 15,511 (range, 41.28 to 98,716). Univariable analyses revealed a significant relationship between the AUC/MIC ratio and mycological response, with the worst response being among patients with lower (≤3,000) AUC/MIC ratios (P=0.005). For patients with Candida parapsilosis, AUC/MIC ratios of ≥285 were predictive of a higher mycological response (P=0.11). Multivariable logistic regression demonstrated the AUC/MIC ratio, APACHE II score, and history of corticosteroid use to be significant independent predictors of a favorable response. PK-PD target attainment analyses suggested that 76.7% and 100% of patients would achieve an AUC/MIC ratio of ≥3,000 for an MIC of 0.03 mg/liter and an AUC/MIC ratio of ≥285 for an MIC of <0.5 mg/liter, respectively. The identification of a lower AUC/MIC ratio target for C. parapsilosis than other Candida species suggests consideration of species-specific echinocandin susceptibility breakpoints and values that are lower than those currently approved by regulatory agencies.
Sigma-Aldrich
Micafungin sodium, ≥97% (HPLC)
Pricing and availability is not currently available.