Merck
  • Lipoxygenase-mediated generation of lipid peroxides enhances ferroptosis induced by erastin and RSL3.
Lipoxygenase-mediated generation of lipid peroxides enhances ferroptosis induced by erastin and RSL3.
Cancer science (2017-08-25)
Ryosuke Shintoku, Yuta Takigawa, Keiichi Yamada, Chisato Kubota, Yuhei Yoshimoto, Toshiyuki Takeuchi, Ichiro Koshiishi, Seiji Torii
ABSTRACT
In cancer cells the small compounds erastin and RSL3 promote a novel type of cell death called ferroptosis, which requires iron-dependent accumulation of lipid reactive oxygen species. Here we assessed the contribution of lipid peroxidation activity of lipoxygenases (LOX) to ferroptosis in oncogenic Ras-expressing cancer cells. Several 12/15-LOX inhibitors prevented cell death induced by erastin and RSL3. Furthermore, siRNA-mediated silencing of ALOX15 significantly decreased both erastin-induced and RSL3-induced ferroptotic cell death, whereas exogenous overexpression of ALOX15 enhanced the effect of these compounds. Immunofluorescence analyses revealed that the ALOX15 protein consistently localizes to cell membrane during the course of ferroptosis. Importantly, treatments of cells with ALOX15-activating compounds accelerated cell death at low, but not high doses of erastin and RSL3. These observations suggest that tumor ferroptosis is promoted by LOX-catalyzed lipid hydroperoxide generation in cellular membranes.
Sigma-Aldrich
1S,3R-RSL 3, ≥98% (HPLC)
Pricing and availability is not currently available.