Merck
  • Removal of toxic Co-EDTA complex by a halophilic solar-salt-pan isolate Pseudomonas aeruginosa SPB-1.
Removal of toxic Co-EDTA complex by a halophilic solar-salt-pan isolate Pseudomonas aeruginosa SPB-1.
Chemosphere (2013-11-05)
A Paraneeiswaran, Sudhir K Shukla, T Subba Rao, K Prashanth
ABSTRACT
In this study, a promising bioremediation approach was developed to remove [Co(III)-EDTA](-) complex that is generated during the waste management process. Though several studies have been reported on bioremediation of cobalt, the removal of [Co(III)-EDTA](-) complex has not been tested. A [Co(III)-EDTA](-) resistant bacterium, Pseudomonas aeruginosa SPB-1 was isolated from the solar-salt-pan and physical parameters were optimized for its growth. The various studies showed that the removal of [Co(III)-EDTA](-) from the bulk liquid was due to the adsorption of the complex by the biomass. Using absorption/desorption isotherm over a range of pH (1-8), the maximum adsorption of [Co(III)-EDTA](-) was found to be at pH 7.0 and maximum desorption from the biomass occurred at pH 1.0, thus rendering an ion exchange property to P. aeruginosa SPB-1 biomass. P. aeruginosa SPB-1 biomass could be used as bio-resin that showed 80.4±3.27% adsorption capacity up to fourth cycle and the biomass was viable till the ninth cycle with 10.5±7.3% adsorption. Radiation tolerance potential i.e. D10 value for the strain was found to be ~300 Gy, which suggests the potential use of the bacterium in bioremediation of moderately active nuclear waste.
Product Description
Supelco
SPB®-1 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm
Pricing and availability is not currently available.