Skip to Content
Merck
TSKgel® Octadecyl-NPR HPLC Column phase C18 (octadecyl), L × I.D. 3.5 cm × 4.6 mm, 2.5 μm particle size

TSKgel® Octadecyl-NPR (2.5 µm) HPLC Columns

TSKgel® Octadecyl-NPR HPLC Column phase C18 (octadecyl), L × I.D. 3.5 cm × 4.6 mm, 2.5 μm particle size
Synonym(s):
TSKgel® Reversed Phase HPLC Column (Polymer-based)

Products

Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1